missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ G6000PW HPLC Column, 17 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ G6000PW SEC columns can be used for the analysis of water-soluble polymers up to 8,000,000 Da molecular weight.
$3218.00 - $5261.80
Specifications
Diameter (Metric) | 7.5 mm |
---|---|
Flow Rate | 1.2 mL/min. |
Particle Size | 17 μm |
pH | 2.0 - 12 |
Pore Size | >1000 Å |
Description
Order Info
Shipped in water.
Specifications
7.5 mm | |
17 μm | |
>1000 Å | |
80°C | |
Hydroxylated Methacrylate | |
Polymer Particle Platform |
1.2 mL/min. | |
2.0 - 12 | |
≤20% organic solvent | |
L38, L39 | |
TSKgel | |
Size Exclusion |
TSKgel is a registered trademark of Tosoh Corporation