missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ G6000PW HPLC Column, 17 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ G6000PW SEC columns can be used for the analysis of water-soluble polymers up to 8,000,000 Da molecular weight.
Supplier: Merck Emd Millipore 805109
Description
Order Info
Shipped in water.
Specifications
7.5 mm | |
60 cm | |
17 μm | |
>1000 Å | |
80°C | |
Hydroxylated Methacrylate | |
Polymer Particle Platform | |
Stainless Steel Hardware |
1.2 mL/min. | |
20 bar | |
2.0 - 12 | |
≤20% organic solvent | |
L38, L39 | |
TSKgel | |
Size Exclusion |
TSKgel is a registered trademark of Tosoh Corporation