Learn More
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls provide results within 3 minutes
$56.24 - $195.95
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Type | Price | Quantity | ||||
361045554
|
Innovatek
419110562 |
50 Tests/Pk. | Latex Test Kit |
Pack of 50 Each for $138.32
|
|
||||
361045555
|
Innovatek
419110563 |
100 Tests/Pk. | Latex Test Kit |
Pack of 100 Each for $195.95
|
|
||||
35-998
|
Innovatek
419110541 |
1 mL | Negative Control |
Each for $56.24
|
|
||||
35-997
|
Innovatek
419110521 |
1 mL | Positive Control |
Each for $56.24
|
|
||||
Description
- Fast 2 to 3 minute rotation
- Complete kit includes all reagents and controls
- Disposable test cards
- 6 to 12 months of shelf life
- Sensitive and specific
Specifications
Rheumatoid Factor (RF) | |
≈3 min. |