Learn More
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls provide results within 3 minutes
Supplier:  Innovatek 419110563
 View more versions of this product
Catalog No. 361045555
Only null left
Explore available promotions
Description
Description
  • Fast 2 to 3 minute rotation
  • Complete kit includes all reagents and controls
  • Disposable test cards
  • 6 to 12 months of shelf life
  • Sensitive and specific
Specifications
Specifications
Rheumatoid Factor (RF)
Latex Test Kit
100 Tests/Pk.
≈3 min.
Product Suggestions
Product Suggestions