Learn More
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls
Innovatek Rheumatoid Factor (RF) Latex Test Kits and Controls provide results within 3 minutes
Supplier: Innovatek 419110562
Description
- Fast 2 to 3 minute rotation
- Complete kit includes all reagents and controls
- Disposable test cards
- 6 to 12 months of shelf life
- Sensitive and specific
Specifications
Rheumatoid Factor (RF) | |
Latex Test Kit | |
50 Tests/Pk. | |
≈3 min. |