Learn More
ABS Premium Laboratory Refrigerator
Delivering superior cooling to laboratory environments.
$16695.77 - $19391.75
Specifications
Type Laboratory Refrigerator
Temperature Range 1°C to 10°C
Cabinet Material Scratch-resistant Painted Steel
Exterior Finish Powder-coated Steel
Certifications/Compliance ETL Listed, ENERGYSTAR
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (English) Door Style Price Quantity  
Catalog Number Mfr. No. Capacity (English) Door Style Price Quantity  
361058604
View Documents
ABS
ABTHCPL23
23 cu. ft. Glass Swing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
361058606
View Documents
ABS
ABTHCPL49
49 cu. ft. Glass Swing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
361058607
View Documents
ABS
ABTHCSPL23
23 cu. ft. Solid Swing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
361058609
View Documents
ABS
ABTHCSPL49
49 cu. ft. Solid Swing N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
Engineered with variable speed compressors (VSCs), these units feature ultra-quiet operation and significant energy savings. VSCs optimize energy consumption by self-adjusting to cooling demands. These compressors also deliver enhanced system performance and provide a longer lifespan than other compressor variations.
  • Ultra-quiet variable speed compressors
  • Superior energy efficiency
  • Microprocessor temperature control
  • Full array of alarms
  • Natural hydrocarbon refrigerants
Enjoy the uniformity and speed of microprocessor temperature control and a full array of alarms to safeguard your products. Unit controllers also come with a battery backup to keep your data safe. Upgrade your laboratory environment with these premium refrigerators and welcome energy savings, noise reduction, smooth operation, and improved system performance to your work environment.
Specifications
Specifications
Laboratory Refrigerator
Scratch-resistant Painted Steel
ETL Listed, ENERGYSTAR
Built-in Temperature and Alarm Event Logging, USB Export
NEMA 5-15
1°C to 10°C
Powder-coated Steel
Upright
Variable Speed
115 V
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'