Learn More
ABS Premium Laboratory Refrigerator
Delivering superior cooling to laboratory environments.
Supplier:  ABS ABTHCPL23
 View more versions of this product
Catalog No. 361058604
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Engineered with variable speed compressors (VSCs), these units feature ultra-quiet operation and significant energy savings. VSCs optimize energy consumption by self-adjusting to cooling demands. These compressors also deliver enhanced system performance and provide a longer lifespan than other compressor variations.
  • Ultra-quiet variable speed compressors
  • Superior energy efficiency
  • Microprocessor temperature control
  • Full array of alarms
  • Natural hydrocarbon refrigerants
Enjoy the uniformity and speed of microprocessor temperature control and a full array of alarms to safeguard your products. Unit controllers also come with a battery backup to keep your data safe. Upgrade your laboratory environment with these premium refrigerators and welcome energy savings, noise reduction, smooth operation, and improved system performance to your work environment.
Specifications
Specifications
Laboratory Refrigerator
651 L
Glass Swing
5
Powder-coated Steel
Upright
Variable Speed
115 V
23 cu. ft.
1°C to 10°C
1
Scratch-resistant Painted Steel
ETL Listed, ENERGYSTAR
Built-in Temperature and Alarm Event Logging, USB Export
NEMA 5-15
Product Suggestions
Product Suggestions