missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt
Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt is a CAT 2 AR/FR, NFPA™ 2112 compliant tee featuring a heat seal Bulwark™ triangle centered at the neck to show the wearer has on an FR undergarment.
$97.31 - $103.38
Specifications
Gender | Men's |
---|---|
Material | FR Cotton |
Color | Navy |
Neck Style | Hemmed Crew |
Sleeve Style | Long Sleeve |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
17-204-057
|
Bulwark™
SET2NVRGS |
Small |
Each for $103.38
|
|
|||||
17-204-205
|
Bulwark™
SET2NVRGM |
Medium |
Each for $103.38
|
|
|||||
17-204-009
|
Bulwark™
SET2NVRGL |
Large |
Each for $103.38
|
|
|||||
17-103-749
|
Bulwark™
SET2NVRGXL |
X-Large |
Each for $97.31
|
|
|||||
17-203-997
|
Bulwark™
SET2NVRGXXL |
2X-Large |
Each for $103.38
|
|
|||||
17-103-750
|
Bulwark™
SET2NVRG3XL |
3X-Large |
Each for $97.31
|
|
|||||
17-204-006
|
Bulwark™
SET2NVRG4XL |
4X-Large |
Each for $103.38
|
|
|||||
17-204-013
|
Bulwark™
SET2NVLNL |
Large (Long) |
Each for $103.38
|
|
|||||
17-204-341
|
Bulwark™
SET2NVLNXL |
X-Large (Long) |
Each for $103.38
|
|
|||||
17-204-063
|
Bulwark™
SET2NVLNXXL |
2X-Large (Long) |
Each for $103.38
|
|
|||||
17-204-333
|
Bulwark™
SET2NVLN3XL |
3X-Large (Long) |
Each for $103.38
|
|
|||||
17-204-095
|
Bulwark™
SET2NVLN4XL |
4X-Large (Long) |
Each for $103.38
|
|
|||||
Description
- Extra comfortable fabric
- Underarm gusset for improved breathability
- Rib-knit crewneck collar
- Single spade chest pocket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m2) Interlock
- Blend: 100% Cotton
- Country of Origin: Imported, Made in USA Fabric
Specifications
Men's | |
Navy | |
Long Sleeve | |
1 | |
8.9 |
FR Cotton | |
Hemmed Crew | |
Reusable | |
Category 2 |