missing translation for 'onlineSavingsMsg'
Learn More

Bulwark™ Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt

Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt is a CAT 2 AR/FR, NFPA™ 2112 compliant tee featuring a heat seal Bulwark™ triangle centered at the neck to show the wearer has on an FR undergarment.

Supplier:  Bulwark™ SET2NVRGS

 View more versions of this product

Catalog No. 17-204-057


Only null left
Explore available promotions

Description

Description

  • Extra comfortable fabric
  • Underarm gusset for improved breathability
  • Rib-knit crewneck collar
  • Single spade chest pocket
  • Hazard: Flash Fire, Arc Flash
  • Wash Care: Home Wash
  • Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m2) Interlock
  • Blend: 100% Cotton
  • Country of Origin: Imported, Made in USA Fabric
Specifications

Specifications

Small
FR Cotton
Hemmed Crew
Reusable
Category 2
ASTM F1506, NFPA 2112, NFPA 70E
Men's
Navy
Long Sleeve
1
8.9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions