missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bulwark™ Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt
Men's Lightweight Navy Long Sleeve Flame Resistant T-shirt is a CAT 2 AR/FR, NFPA™ 2112 compliant tee featuring a heat seal Bulwark™ triangle centered at the neck to show the wearer has on an FR undergarment.
Supplier: Bulwark™ SET2NVRGXXL
Description
- Extra comfortable fabric
- Underarm gusset for improved breathability
- Rib-knit crewneck collar
- Single spade chest pocket
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: EXCEL FR™ Flame resistant, 7 oz. (235 g/m2) Interlock
- Blend: 100% Cotton
- Country of Origin: Imported, Made in USA Fabric
Specifications
2X-Large | |
FR Cotton | |
Hemmed Crew | |
Reusable | |
Category 2 | |
ASTM F1506, NFPA 2112, NFPA 70E |
Men's | |
Navy | |
Long Sleeve | |
1 | |
8.9 |