missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, 99.8%, ACS DEA List I, Reagents
$113.23 - $12851.10
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
361059157
|
Labchem Inc
C1217500100C2 |
100 g |
Each for $368.23
|
|
|||||
361059156
|
Labchem Inc
C121750012A5 |
12 kg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
361059158
|
Labchem Inc
C121750025C1 |
25 g |
Each for $113.23
|
|
|||||
361059161
|
Labchem Inc
C12175002KG |
2 kg |
Each for $2,897.13
|
|
|||||
361059163
|
Labchem Inc
C1217500500C2 |
500 g |
Each for $978.37
|
|
|||||
Description
- Iodine, ACS Grade
- Brand: Reagents
- DEA List I
- Appearance: Violet-black Crystals
- Molecular Formula I2
- MW (g/mol): 253.8089
- Assay >= 99.8 %
- Chlorine and Bromine (as Cl) <= 0.005 %
- Nonvolatile matter <= 0.01 %
Chemical Identifiers
253.81 |
Specifications
Crystals and/or Chunks | |
253.81 | |
ACS |
100 g | |
99.80% |