missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, 99.8%, ACS DEA List I, Reagents
Supplier: Labchem Inc C121750025C1
Alert:
DEA LIST I
Description
- Iodine, ACS Grade
- Brand: Reagents
- DEA List I
- Appearance: Violet-black Crystals
- Molecular Formula I2
- MW (g/mol): 253.8089
- Assay >= 99.8 %
- Chlorine and Bromine (as Cl) <= 0.005 %
- Nonvolatile matter <= 0.01 %
Specifications
Crystals and/or Chunks | |
253.81 | |
ACS |
25 g | |
99.80% |
Chemical Identifiers
253.81 |