missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 99.8%, ACS DEA List I, Reagents

Supplier:  Labchem Inc C121750012A5

 View more versions of this product

Catalog No. 361059156

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Alert:

DEA LIST I


Description

Description

  • Iodine, ACS Grade
  • Brand: Reagents
  • DEA List I
  • Appearance: Violet-black Crystals
  • Molecular Formula I2
  • MW (g/mol): 253.8089
  • Assay >= 99.8 %
  • Chlorine and Bromine (as Cl) <= 0.005 %
  • Nonvolatile matter <= 0.01 %
Specifications

Specifications

Crystals and/or Chunks
253.81
ACS
12 kg
99.80%

Chemical Identifiers

253.81
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions