missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.5 N, Aqueous Solution, Spectrum™ Chemical

$104.82 - $1,946.09

Chemical Identifiers

CAS 7647-01-0
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610875
View Documents
Spectrum Chemical Mfg Cor
H1301LT
1 L Poly Bottle N/A
Only null left
 
18610876
View Documents
Spectrum Chemical Mfg Cor
H1304LTPL
4 L Poly Bottle N/A
Only null left
 
18610877
View Documents
Spectrum Chemical Mfg Cor
H13020LT
20 L Cubitainer N/A
Only null left
 
18610878
View Documents
Spectrum Chemical Mfg Cor
H130200LTBL
200 L Poly Drum N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7647-01-0

Specifications

7647-01-0,7732-18-5
0.5 N ± 0.005 N
Poly Bottle
1 L
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.