missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric Acid, 0.5 N, Aqueous Solution, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor H13020LT
Spécifications
7647-01-0,7732-18-5 | |
0.5 N ± 0.005 N | |
Cubitainer | |
20 L |
Identifiants chimiques
7647-01-0 |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.