missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid, 0.5 N, Aqueous Solution, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor H1304LTPL

 View more versions of this product

Catalog No. 18610876


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7647-01-0,7732-18-5
0.5 N ± 0.005 N
Poly Bottle
4 L

Chemical Identifiers

7647-01-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.