missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Zenix™ SEC-300 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel
Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.
Supplier: MilliporeSigma™ Supelco™ Z777027
Description
Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.Specifications
4.6 mm | |
3 μm | |
300 Å | |
Zenix | |
Size Exclusion | |
SEC HPLC |
25 cm | |
2.0 to 8.5 | |
Hydrophilic Polymer | |
Hydrophilic Polymer Phase/Spherical Silica | |
Stainless Steel |
Zenix is a registered trademark of Sepax Technologies