missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Zenix™ SEC-100 Gel Filtration HPLC Column, 3 μm Particle Size, Stainless Steel
Like their 5 micron siblings, 3 micron Zenix™ SEC columns feature a hydrophilic surface layer to prevent the sample from interacting with the underlying silica.
$2935.24 - $3336.77
Specifications
Length (Metric) | 30 cm |
---|---|
Particle Size | 3 μm |
pH | 2.0 to 8.5 |
Pore Size | 100 Å |
Stationary Phase | Hydrophilic Polymer |
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
111005955
|
MilliporeSigma™ Supelco™
Z777002 |
1 mm |
Each for $3,336.77
|
|
|||||
111005956
|
MilliporeSigma™ Supelco™
Z777004 |
2.1 mm |
Each for $3,336.77
|
|
|||||
111006060
|
MilliporeSigma™ Supelco™
Z777006 |
4.6 mm |
Each for $2,935.24
|
|
|||||
111005957
|
MilliporeSigma™ Supelco™
Z777008 |
7.8 mm |
Each for $3,336.77
|
|
|||||
Description
Select 100, 300 or 500 Å pores based on the size of your sample molecules, whether you are analyzing peptides, proteins, mAbs, ADCs, fusion proteins, etc. in quality control or research.Specifications
30 cm | |
2.0 to 8.5 | |
Hydrophilic Polymer | |
Hydrophilic Polymer Phase/Spherical Silica | |
Stainless Steel |
3 μm | |
100 Å | |
Zenix | |
Size Exclusion | |
SEC HPLC |
Zenix is a registered trademark of Sepax Technologies