missing translation for 'onlineSavingsMsg'
Learn More

Watson Gloves Yellow Tail Welding Gloves

Yellow Tail Welding Gloves have a full-grain deerskin leather palm, split cowhide back, foam lined back and gunn cut design with wing thumb.

$214.48 - $214.48

Specifications

Chemotherapeutic Rated No
Cuff Material Leather Gore
Color Blue, Brown
Grade Premium
Hand Ambidextrous
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
17-099-927
View Documents
Watson Gloves
2752M
Medium
Case of 6 Pair for $214.48
Only null left
 
17-099-931
View Documents
Watson Gloves
2752L
Large
Case of 6 Pair for $214.48
Only null left
 
17-099-864
View Documents
Watson Gloves
2752X
X-Large
Case of 6 Pair for $214.48
Only null left
 
Description

Description

  • Full-grain deerskin leather palm
  • Split cowhide back
  • Foam lined back
  • Gunn cut design with wing thumb
  • Reinforced welted seams
  • Gauntlet style cuff
Specifications

Specifications

No
Blue, Brown
Ambidextrous
Welding Gloves
Welding
Blue/Gray
Full Grain Deerskin
Wing
5 in.
13 cm
Leather Gore
Premium
Deerskin Leather, Split Cowhide Leather
Gunn
Men's
Gauntlet
Heat wave
12 in.
30 cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'