missing translation for 'onlineSavingsMsg'
Learn More

Urea, Crystal, ACS, 99.0-100.5%, Spectrum™ Chemical

$421.67 - $10494.20

Chemical Identifiers

CAS 57-13-6
Molecular Formula CH4N2O
Molecular Weight (g/mol) 60.06
InChI Key XSQUKJJJFZCRTK-UHFFFAOYSA-N
IUPAC Name urea
SMILES NC(N)=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614626
View Documents
Spectrum Chemical Mfg Cor
U1015500GM
500 g Poly Bottle
Each for $421.67
Only null left
 
18614625
View Documents
Spectrum Chemical Mfg Cor
U101525KG
2.5 kg Poly Pail
Each for $1,114.13
Only null left
 
18614624
View Documents
Spectrum Chemical Mfg Cor
U101512KG
12 kg Poly Pail
Each for $3,455.83
Only null left
 
18614627
View Documents
Spectrum Chemical Mfg Cor
U101550KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

57-13-6
60.06
urea
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
NC(N)=O

Specifications

57-13-6
132° to 135°C
Poly Bottle
500 g
NC(N)=O
60.06
ACS
1
0.0001
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
99.0 to 100.5%
0.0001
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.