missing translation for 'onlineSavingsMsg'
Learn More

Urea, Crystal, ACS, 99.0-100.5%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor U101512KG

 View more versions of this product

Catalog No. 18614624


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

57-13-6
132° to 135°C
Poly Pail
12 kg
NC(N)=O
60.06
ACS
1
0.0001
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
urea
99.0 to 100.5%
0.0001

Chemical Identifiers

57-13-6
60.06
urea
CH4N2O
XSQUKJJJFZCRTK-UHFFFAOYSA-N
NC(N)=O
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.