Learn More
DuPont™ Tychem™ 6000 Coverall with Attached Hood, Socks, Butyl Gloves
Tychem™ 6000 Coveralls provide protection from chemical warfare agents and toxic industrial chemicals.
$2034.45 - $2716.61
Specifications
Color Gray
Material Tychem
Ankle Style Boot Overflaps
Cuff Style Attached Gloves
Description Packaged Individually
View More Specs
Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
18888788
View Documents
DuPont™
TF186TGY4X0006PI
4X-Large
Case of 6 for $2,515.35
Only null left
18-888-789
View Documents
DuPont™
TF186TGY5X0006PI
5X-Large
Case of 6 Each for $2,716.61
Only null left
18-888-792
View Documents
DuPont™
TF186TGYSM0006PI
Small
Case of 6 Each for $2,034.45
Only null left
18888786
View Documents
DuPont™
TF186TGY2X0006PI
2X-Large
Case of 6 Each for $2,156.54
Only null left
18888787
View Documents
DuPont™
TF186TGY3X0006PI
3X-Large
Case of 6 Each for $2,329.08
Only null left
18-888-790
View Documents
DuPont™
TF186TGYLG0006PI
Large
Case of 6 Each for $2,034.45
Only null left
18-888-791
View Documents
DuPont™
TF186TGYMD0006PI
Medium
Case of 6 Each for $2,034.45
Only null left
18-888-793
View Documents
DuPont™
TF186TGYXL0006PI
X-Large
Case of 6 Each for $2,034.45
Only null left
Description
Description
Tychem™ 6000 coveralls are made of a proprietary barrier film laminated to a Tyvek™ substrate. Tychem™ 6000 fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Used by military personnel and first responders for chemical warfare situations
  • Available in low-visibility gray fabric for discretionary purpose
Specifications
Specifications
Gray
Boot Overflaps
Packaged Individually
Unisex
Hooded
6/Pk.
Taped
Tychem
Attached Gloves
Single-use
Yes
None
Liquid
Non-sterile
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'