Learn More
DuPont™ Tychem™ 6000 Coverall with Attached Hood, Socks, Butyl Gloves
Tychem™ 6000 Coveralls provide protection from chemical warfare agents and toxic industrial chemicals.
Supplier:  DuPont™ TF186TGYLG0006PI
 View more versions of this product
Catalog No. 18-888-790
Only null left
Explore available promotions
Description
Description
Tychem™ 6000 coveralls are made of a proprietary barrier film laminated to a Tyvek™ substrate. Tychem™ 6000 fabrics are tested by a third party laboratory to assess its chemical permeation resistance.
  • Provides protection against over 180 chemical challenges for at least 30 minutes
  • Used by military personnel and first responders for chemical warfare situations
  • Available in low-visibility gray fabric for discretionary purpose
Specifications
Specifications
Gray
Large
Attached Gloves
Single-use
Yes
None
Liquid
Non-sterile
Tychem
Boot Overflaps
Packaged Individually
Unisex
Hooded
6/Pk.
Taped
Product Suggestions
Product Suggestions