Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Viton Showa Best™ 890 Outer Gloves
Tychem™ 10000 offers a high level of protection against toxic, corrosive gases and liquid chemicals
$2004.22 - $2385.15
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
18-888-858
|
DuPont™
TK555TLY2X00015V |
Case for $2,111.26
|
|
||||||
18-888-862
|
DuPont™
TK555TLY3X00015V |
Case for $2,111.26
|
|
||||||
18-888-865
|
DuPont™
TK555TLY4X00015V |
Case for $2,385.15
|
|
||||||
18-888-868
|
DuPont™
TK555TLYLG00015V |
Case for $2,004.22
|
|
||||||
18888872
|
DuPont™
TK555TLYMD00015V |
Case for $2,004.22
|
|
||||||
18888875
|
DuPont™
TK555TLYXL00015V |
Case for $2,004.22
|
|
||||||
Description
Tychem™ 10000 has excellent chemical barrier properties and high strength for enhanced durability. Developed for protection against toxic, corrosive gases and liquid chemicals, this high-performance chemical barrier fabric is suited for industrial, HAZMAT, and domestic preparedness applications. Tychem™ 10000 garments are available to meet NFPA 1991 and NFPA 1994 standards.
- Provides at least 30 minutes of protection against over 320 chemical challenges
- Chosen by HAZMAT responders worldwide
- Available in high-visibility lime yellow
- Garments include fully encapsulated, vapor protective Level A suit and liquid-splash protective Level B suits as well as coveralls.
Specifications
Tychem |