Learn More
DuPont™ Tychem™ 10000 Encapsulated Level A Suit with Viton Showa Best™ 890 Outer Gloves
Tychem™ 10000 offers a high level of protection against toxic, corrosive gases and liquid chemicals
Supplier:  DuPont™ TK555TLYLG00015V
 View more versions of this product
Catalog No. 18-888-868
Only null left
Explore available promotions
Description
Description
Tychem™ 10000 has excellent chemical barrier properties and high strength for enhanced durability. Developed for protection against toxic, corrosive gases and liquid chemicals, this high-performance chemical barrier fabric is suited for industrial, HAZMAT, and domestic preparedness applications. Tychem™ 10000 garments are available to meet NFPA 1991 and NFPA 1994 standards.
  • Provides at least 30 minutes of protection against over 320 chemical challenges
  • Chosen by HAZMAT responders worldwide
  • Available in high-visibility lime yellow
  • Garments include fully encapsulated, vapor protective Level A suit and liquid-splash protective Level B suits as well as coveralls.
Specifications
Specifications
Tychem
Product Suggestions
Product Suggestions