missing translation for 'onlineSavingsMsg'
Learn More

Wayne Safety™ Tuff Blue Nitrile Gloves

These powder-free nitrile gloves are made of 100% nitrile without fillers or natural rubber latex. Ideal for the healthcare setting.

$8.64 - $8.88

Specifications

Certifications/Compliance CFIA Approved, Health Canada Class 2 Medical Device License
Certifications/Compliance (Europe) EN374, EN455
Applications Healthcare
Color Blue
Disposable Single-use
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
29-635-1434
View Documents
Wayne Safety™
15I700PFS
Small
Pack of 100 Each for $8.72
Case of 10 Pack for $87.20
Only null left
 
29-635-1435
View Documents
Wayne Safety™
15I700PFM
Medium
Pack of 100 Each for $8.72
Case of 10 Pack for $87.20
Only null left
 
29-635-1436
View Documents
Wayne Safety™
15I700PFL
Large
Pack of 100 Each for $8.64
Case of 10 Pack for $86.40
Only null left
 
29-635-1437
View Documents
Wayne Safety™
15I700PFXL
X-Large
Pack of 100 Each for $8.88
Case of 10 Pack for $88.80
Only null left
 
Description

Description

  • Puncture and tear-resistant for prolonged usage
  • Ideal sensitivity
  • Better protection against oil, grease and organic solvents
  • Approved for use to protect against Fentanyl Citrate permeation and other healthcare applications
Specifications

Specifications

CFIA Approved, Health Canada Class 2 Medical Device License
Healthcare
Single-use
Yes
Powder Free
Non-sterile
100 Gloves/Box
EN374, EN455
Blue
Smooth
Nitrile
Nitrile Examination Glove
No
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'