missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ SuperHZM-H Guard Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ SuperHZM-H Mixed-Bed Columns are packed with rigid, porous, polymer beads each having a varying mean pore diameter. They are used for the non-aqueous separation of organic-soluble polymers with high molecular weights of less than 400,000,000 Da.
$1208.86 - $1208.86
Specifications
Particle Size | 10 μm |
---|---|
Diameter (Metric) Inner | 4.6 mm |
Flow Rate | 0.4 mL/min |
Stationary Phase | Polystyrene-divinylbenzene |
Temperature (Metric) Maximum | 80°C |
Description
- THF shipping solvent may be replaced by a limited number of organic solvents
Specifications
10 μm | |
0.4 mL/min | |
80°C | |
Size Exclusion (SEC) |
4.6 mm | |
Polystyrene-divinylbenzene | |
Stainless Steel | |
TSKgel |
TSKgel is a registered trademark of Tosoh Corporation