missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKgel™ ODS-120A HPLC Column, 10 μm Particle Size, Sold by MilliporeSigma™ Supelco™
TSKgel™ ODS-120A columns contain 150 Å pore size silica particles. The exclusion limit of 10,000 Da makes TSKgel™ ODS-120A columns a good choice for the reversed phase chromatography of peptides, small proteins, and environmental samples such as poly-aromatic hydrocarbons.
$3377.54 - $7924.58
Specifications
Length (Metric) | 30 cm |
---|---|
Particle Size | 10 μm |
pH | 2.0 - 7.5 |
Pore Size | 150 Å |
Solvents | 0 to 100% organic solvent |
Description
The 'A' in the product description signifies that the material is not endcapped. For charged samples, the endcapped TSKgel™ ODS-120T column is a more suitable alternative.Order Info
Shipped in 70% methanol - 30% water.
Specifications
30 cm | |
2.0 - 7.5 | |
0 to 100% organic solvent | |
L1 | |
TSKgel | |
20% | |
Reversed Phase |
10 μm | |
150 Å | |
0°C to 45°C | |
C18 (octadecyl) | |
Spherical Silica | |
No |
TSKgel is a registered trademark of Tosoh Corporation