missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

$4169.59 - $6139.88

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
2004534
View Documents
Avantor J.T.Baker
946801
500 mL Narrow Mouth Amber Glass Bottle
Case of 12 Each for $4,169.59
Only null left
 
2004533
View Documents
Avantor J.T.Baker
946803
4 L Narrow Mouth Amber Glass Bottle
Case of 4 Each for $6,139.88
Only null left
 
Description

Description

Specifications

Chemical Identifiers

102-71-6
149.19
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
CHEBI:28621
C(CO)N(CCO)CCO
C6H15NO3
GSEJCLTVZPLZKY-UHFFFAOYSA-N
7618
2-[bis(2-hydroxyethyl)amino]ethanol

Specifications

102-71-6
Narrow Mouth Amber Glass Bottle
500 mL
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
C(CO)N(CCO)CCO
149.19
CHEBI:28621
BAKER ANALYZED™ Reagent
179.00°C
C6H15NO3
1L = 1.13kg
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanol
7618
149.19
Triethanolamine
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'