missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, BAKER ANALYZED™ Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Supplier:  Avantor J.T.Baker 946803

 View more versions of this product

Catalog No. 2004533


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Triethanolamine
179.00°C
C6H15NO3
1L = 1.13kg
GSEJCLTVZPLZKY-UHFFFAOYSA-N
2-[bis(2-hydroxyethyl)amino]ethanol
7618
149.19
102-71-6
Narrow Mouth Amber Glass Bottle
4 L
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
C(CO)N(CCO)CCO
149.19
CHEBI:28621
BAKER ANALYZED™ Reagent

Chemical Identifiers

102-71-6
149.19
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
CHEBI:28621
C(CO)N(CCO)CCO
C6H15NO3
GSEJCLTVZPLZKY-UHFFFAOYSA-N
7618
2-[bis(2-hydroxyethyl)amino]ethanol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions