Learn More
Corepoint™ Scientific Touchscreen Manual Defrost Laboratory and Medical Freezer with Hydrocarbon Refrigerants
This touchscreen manual defrost freezer for medical and laboratory purposes offer professional cold storage for scientific and industrial settings.
Supplier:  Corepoint™ Scientific NSPF201WWW0MHCTS
 View more versions of this product
Catalog No. 361058675
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
Units come equipped with:
  • Microprocessor temperature control
  • Array of temperature alarms
  • Probe access
  • Keyed door locks
  • Natural hydrocarbon refrigerants
  • Our freezers showcase elite features including state-of-the-art touchscreen controllers, advanced alarms and data logging capabilities and unparalleled temperature control.
  • All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Specifications
Specifications
Laboratory Freezer
481 L
Solid Swing
Scratch-resistant Painted Steel
6
Upright
NEMA 5-15
H-drive
17 cu. ft.
-15°C to -25°C
1
Painted Steel
cULus
Internal Data Logger USB
115 V
Product Suggestions
Product Suggestions