missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Purified, Reagents

Toluene, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

$109.77 - $6878.53

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym Methylbenzene, Phenylmethane, Toluol
IUPAC Name toluene
SMILES CC1=CC=CC=C1
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-336-927
View Documents
Labchem Inc
C2341000200E
200 L
Each for $6,878.53
Only null left
 
01-336-926
View Documents
Labchem Inc
C234100020E6
20 L
Each for $1,026.83
Only null left
 
01-336-925
View Documents
Labchem Inc
C23410004C
4 L
Each for $342.30
Only null left
 
01-336-924
View Documents
Labchem Inc
C2341000500C
500 mL
Each for $109.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

108-88-3
92.14
Methylbenzene, Phenylmethane, Toluol
CC1=CC=CC=C1
C7H8
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene

Specifications

-95°C
Colorless
200 L
Liquid
C7H8
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
Pure
108-88-3
111°C
Steel Drum
0.866
Methylbenzene, Phenylmethane, Toluol
CC1=CC=CC=C1
92.14
Toluene, Purified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'