missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Toluene, Purified, Reagents
Toluene, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2341000500C
Specifications
Toluene, Purified | |
108-88-3 | |
111°C | |
Glass Bottle | |
0.866 | |
Methylbenzene, Phenylmethane, Toluol | |
CC1=CC=CC=C1 | |
92.14 |
-95°C | |
Colorless | |
500 mL | |
Liquid | |
C7H8 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
Pure |
Chemical Identifiers
108-88-3 | |
92.14 | |
Methylbenzene, Phenylmethane, Toluol | |
CC1=CC=CC=C1 |
C7H8 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene |