missing translation for 'onlineSavingsMsg'
Learn More

Synaptoporin Monoclonal Antibody (K49016_14F8), Invitrogen™

Mouse Monoclonal Antibody

$1421.98 - $2134.69

Specifications

Antigen Synaptoporin
Clone K49016_14F8
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA556003
View Documents
Invitrogen™
MA556003
200 μg
Each for $2,134.69
Only null left
 
MA555623
View Documents
Invitrogen™
MA555623
50 μg
Each for $1,421.98
Only null left
 
Description

Description

Sequence of this protein is as follows: TGWHSSGQRY LSDPMEKHSS

Synaptoporin Monoclonal antibody specifically detects Synaptoporin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Synaptoporin
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide
1500003F20Rik; SPO; Synaptoporin; synaptorin; SYNPR
Human SYNPR peptide (immunogen range is 201-220).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
SYNPR
K49016_14F8
Western Blot
Unconjugated
Mouse
Human, Mouse, Rat
P22831, Q8BGN8, Q8TBG9
132204, 66030, 72003
IgG1
Protein A
Antibody
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'