missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Synaptoporin Monoclonal Antibody (K49016_14F8), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555623
Description
Sequence of this protein is as follows: TGWHSSGQRY LSDPMEKHSS
Synaptophysin 2, also known as synaptoporin, is a component of the synaptic vesicle membrane and thought to play an important role in synaptic vesicle trafficking (Dai et al., 2003). It is highly homologous to synaptophysin 1, yet differs in expression. Whereas synaptophysin 1 is ubiquitously expressed, synaptophysin 2 is concentrated in the mossy fiber synapses of the hippocampus (Grabs et al., 1994; Singec et al., 2002).Specifications
Synaptoporin | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
P22831, Q8BGN8, Q8TBG9 | |
SYNPR | |
Human SYNPR peptide (immunogen range is 201-220). | |
50 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Western Blot | |
K49016_14F8 | |
Unconjugated | |
SYNPR | |
1500003F20Rik; SPO; Synaptoporin; synaptorin; SYNPR | |
Mouse | |
Protein A | |
RUO | |
132204, 66030, 72003 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |