missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ IBA-35, X-Heavy Chemical Resistant Butyl Gloves
Extra heavyweight butyl gloves are 35 mil thick and feature a smooth finish to protect against a wide range of hazardous chemicals.
$145.97 - $145.97
Specifications
Certifications/Compliance | ASTM F739-05 |
---|---|
Certifications/Compliance (Europe) | CE |
Color | Black |
Hand | Specific |
Latex Free | Yes |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19321114
|
Guardian™
53501 |
X-Small |
Pair for $145.97
|
|
|||||
19321115
|
Guardian™
53502 |
Small |
Pair for $145.97
|
|
|||||
19321116
|
Guardian™
53503 |
Medium |
Pair for $145.97
|
|
|||||
19321117
|
Guardian™
53504 |
Large |
Pair for $145.97
|
|
|||||
19321118
|
Guardian™
53505 |
X-Large |
Pair for $145.97
|
|
|||||
Description
- Extra heavy butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 35 mil thickness, 14 in. length
- Made in the USA
Specifications
ASTM F739-05 | |
Black | |
Yes | |
Powdered | |
0.88 mm | |
Gauntlet/Rolled | |
Unlined | |
14 in. |
CE | |
Specific | |
Butyl | |
35 mil | |
Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones | |
Smooth | |
ISO 9001 | |
35.5 cm |