missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Guardian™ IBA-35, X-Heavy Chemical Resistant Butyl Gloves
Extra heavyweight butyl gloves are 35 mil thick and feature a smooth finish to protect against a wide range of hazardous chemicals.
Supplier: Guardian™ 53505
Description
- Extra heavy butyl material resists oxidation, ozone corrosion and abrasions
- Reusable and easy to clean
- Fitted hand style with smooth texture
- Protects against wide range of hazardous chemicals
- 35 mil thickness, 14 in. length
- Made in the USA
Specifications
ASTM F739-05 | |
Black | |
Yes | |
Powdered | |
35 mil | |
Permeation resistance to most acids, aldehydes, alcohols, amides, bases, esters and ketones | |
Smooth | |
ISO 9001 | |
14 in. |
CE | |
Specific | |
Butyl | |
X-Large | |
0.88 mm | |
Gauntlet/Rolled | |
Unlined | |
53505 | |
35.5 cm |