missing translation for 'onlineSavingsMsg'
Learn More

Ambion™ Silencer™ Select Pre-Designed siRNA DFS Item

Silencer™ Select Pre-Designed siRNA

$427.00 - $427.00

Specifications

RNAi Type siRNA
Final Product Type Silencer™ Select pre-designed siRNA
For Use With (Application) RNAi, epigenetics and non-coding RNA research, RNAi, siRNA
Product Line Silencer™ Select, Ambion™
Product Type Silencer™ Select pre-designed siRNA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
43-907-71 DFS Item
View Documents
Ambion™
4390771
5 nmol
Each for $427.00
Only null left
 
Description

Description

The Silencer™ Select siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. A choice of Silencer™ Select siRNAs are available—Pre-Designed or Validated. Silencer™ Select Pre-Designed siRNAs are guaranteed-to-silence based on their proven design while the Silencer™ Select Validated siRNAs are functionally tested to reduce target gene expression. Sets of Silencer™ Select siRNA Libraries are also available in tubes or plate for high throughput screening. The Silencer™ Select Positive and Negative Control RNAs should be included in every siRNA experiment.

The benefits of using Silencer™ Select siRNAs include:
  • Potent—up to 100-fold more potent than currently available siRNAs and fewer off-target effects
  • Most Specific—LNA™ chemical modifications reduce off-target effects by up to 90%
  • Reliable—demonstrated improvements in consistency and reliability of phenotypic results
  • Guaranteed—100% guaranteed to silence, the best guarantee in the industry

  • Note: Silencer™ Pre-Designed & Validated siRNAs are designed using our first-generation algorithm and DO NOT include any chemical modifications. They are recommended for completing in vitro RNAi studies that were started using Silencer™ siRNAs. For new studies we recommend Silencer Select siRNAs.

    Approximate Turnaround Times*
  • Standard purification: 4 business days
  • HPLC purification: 6 business days (20 and 40 nmol); 15 business days (250 nmol)
  • In Vivo Ready: 15 business days (250 nmol)
  • Specifications

    Specifications

    siRNA
    RNAi, epigenetics and non-coding RNA research, RNAi, siRNA
    Silencer™ Select pre-designed siRNA
    Silencer™ Select pre-designed siRNA
    Silencer™ Select, Ambion™
    Standard purity
    Videos
    SDS
    missing translation for 'documents'

    missing translation for 'documents'

    For Research Use Only. Not for use in diagnostic procedures.