missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ SGE Autosampler Syringes, Cemented Needle (N)

For use with CTC/LEAP PAL GC

Supplier:  MilliporeSigma™ Supelco™ 29625U

 View more versions of this product

Catalog No. 111011370


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Stainless Steel (Tip)
26 G
SGE Autosampler Syringe
1 Ea.
50 mm
CTC, LEAP
Dark Orange
0.47 mm
1
10 μL
Cone
Cemented
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions