missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Versilon™ SE-200 Tubing DFS Item

Phthalate-free chemical inert transfer Versilon™ SE-200 Tubing.

$512.70 - $512.70

Specifications

Autoclavable Not Autoclavable
Material FEP
Overall Length (English) 50 ft.
Overall Length (Metric) 15 m
For Use With Chemical processing, semiconductor, adhesive transfer, fertilizer and pesticide distribution
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Maximum Pressure Thickness (Metric) Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Inner Diameter (Metric) Outer Maximum Pressure Thickness (Metric) Price Quantity  
14-171-124 DFS Item
View Documents
Saint Gobain Life Sciences
AJD00007
3.2 mm 6.35 mm 85 psi 1.6 mm
Pack of 50 Each for $512.70
Only null left
 
Description

Description

Offering flexibility, glass-like clarity and outstanding bend radius, Versilon SE-200 can handle many applications where flexible tubing of the past could not be used. Its FEP inner liner provides the ultimate in chemical resistance and can accommodate a wide variety of fluids, from corrosives to MEK-based solvents. The inert liner limits the potential of fluid contamination during transfer.

  • Crystal clarity
  • Improved flexibility when compared to rigid fluoropolymer tubing
  • Chemically resistant and inert
  • Non-contaminating fluid path
  • Contains no BPA or phthalates

Typical Applications

  • Chemical processing
  • Paint and solvent production and packaging
  • Adhesive transfer lines
  • Semiconductor processing
  • Photographic processing equipment
  • Fertilizer and pesticide distribution
Specifications

Specifications

Not Autoclavable
50 ft.
Chemical processing, semiconductor, adhesive transfer, fertilizer and pesticide distribution
-36°C
250 lb-f/in
Clear
165°F
Tubing
FEP
15 m
-32°F
1 Ea.
Non-sterile
65 (Shore A)
74°C
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'