missing translation for 'onlineSavingsMsg'
Learn More

Polyvinyl Alcohol, USP, Spectrum™ Chemical

P1180, 9002-89-5, (C2H4O)n

$5400.33 - $107157.53

Chemical Identifiers

CAS 9002-89-5
Molecular Formula (C2H4O)n
Molecular Weight (g/mol) 44.05
MDL Number MFCD00081922
InChI Key IMROMDMJAWUWLK-UHFFFAOYSA-N
IUPAC Name ethenol
SMILES OC(-*)C-*
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18605409
View Documents
Spectrum Chemical Mfg Cor
P11801KG
1 kg Poly Bottle
Each for $5,400.33
Only null left
 
18605410
View Documents
Spectrum Chemical Mfg Cor
P118012KGBL
12 kg Poly Pail N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18605413
View Documents
Spectrum Chemical Mfg Cor
P118025KGBL
25 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
18605411
View Documents
Spectrum Chemical Mfg Cor
P118050KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Polyvinyl Alcohol, USP is used to relieve dry, irritated eyes. All Spectrum Chemical USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Chemical Identifiers

9002-89-5
44.05
IMROMDMJAWUWLK-UHFFFAOYSA-N
OC(-*)C-*
(C2H4O)n
MFCD00081922
ethenol

Specifications

5.0 to 8.0
3
(C2H4O)n
1 kg
OC(-*)C-*
44.05
0.01
Poly Bottle
9002-89-5
1
MFCD00081922
IMROMDMJAWUWLK-UHFFFAOYSA-N
ethenol
USP
0.05
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'