missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Polyvinyl Alcohol, USP, Spectrum™ Chemical
P1180, 9002-89-5, (C2H4O)n
Supplier: Spectrum Chemical Mfg Cor P118012KGBL
Description
Spectrum™ Chemical Polyvinyl Alcohol, USP is used to relieve dry, irritated eyes. All Spectrum Chemical USP are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
5.0 to 8.0 | |
3 | |
(C2H4O)n | |
12 kg | |
OC(-*)C-* | |
44.05 | |
0.01 | |
Poly Pail |
9002-89-5 | |
1 | |
MFCD00081922 | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
ethenol | |
USP | |
0.05 |
Chemical Identifiers
9002-89-5 | |
44.05 | |
IMROMDMJAWUWLK-UHFFFAOYSA-N | |
OC(-*)C-* |
(C2H4O)n | |
MFCD00081922 | |
ethenol |