missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Anhydride, 98+% (1,3-Isobenzofurandione), Reagents
Phthalic Anhydride, 98+% (1,3-Isobenzofurandione), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$254.43 - $763.47
Chemical Identifiers
CAS | 85-44-9 |
---|---|
Molecular Formula | C8H4O3 |
Molecular Weight (g/mol) | 148.12 |
InChI Key | LGRFSURHDFAFJT-UHFFFAOYSA-N |
Synonym | 1,3-Isobenzofurandione |
IUPAC Name | 1,3-dihydro-2-benzofuran-1,3-dione |
SMILES | O=C1OC(=O)C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
01-337-131
|
Labchem Inc
C227400025A5 |
2.5 kg |
Each for $763.47
|
|
|||||
01-337-130
|
Labchem Inc
C2274000500A5 |
500 g |
Each for $254.43
|
|
|||||
Chemical Identifiers
85-44-9 | |
148.12 | |
1,3-Isobenzofurandione | |
O=C1OC(=O)C2=CC=CC=C12 |
C8H4O3 | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
1,3-dihydro-2-benzofuran-1,3-dione |
Specifications
131°C | |
284°C | |
C8H4O3 | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
1,3-dihydro-2-benzofuran-1,3-dione | |
Reagent | |
Poly Bottle | |
1.527 |
85-44-9 | |
White | |
1,3-Isobenzofurandione | |
O=C1OC(=O)C2=CC=CC=C12 | |
148.12 | |
2.5 kg | |
Solid | |
Phthalic Anhydride, 98+% (1,3-Isobenzofurandione) |