missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Anhydride, 98+% (1,3-Isobenzofurandione), Reagents
Phthalic Anhydride, 98+% (1,3-Isobenzofurandione), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2274000500A5
Specifications
Phthalic Anhydride, 98+% (1,3-Isobenzofurandione) | |
85-44-9 | |
White | |
1,3-Isobenzofurandione | |
O=C1OC(=O)C2=CC=CC=C12 | |
148.12 | |
500 g | |
Solid |
131°C | |
284°C | |
C8H4O3 | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
1,3-dihydro-2-benzofuran-1,3-dione | |
Reagent | |
Poly Bottle | |
1.527 |
Chemical Identifiers
85-44-9 | |
148.12 | |
1,3-Isobenzofurandione | |
O=C1OC(=O)C2=CC=CC=C12 |
C8H4O3 | |
LGRFSURHDFAFJT-UHFFFAOYSA-N | |
1,3-dihydro-2-benzofuran-1,3-dione |