missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, White, Powder, 98.0-101.0%, Spectrum™ Chemical
$547.00 - $25710.87
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18-612-279
|
Spectrum Chemical Mfg Cor
P1591125GM |
125 g | Poly Pail |
Each for $547.00
|
|
||||
18-612-281
|
Spectrum Chemical Mfg Cor
P1591500GM |
500 g | Poly Pail |
Each for $1,417.60
|
|
||||
18-612-278
|
Spectrum Chemical Mfg Cor
P159125KG |
2.5 kg | Poly Pail |
Each for $4,958.93
|
|
||||
18-612-280
|
Spectrum Chemical Mfg Cor
P159112KG |
12 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
18-612-282
|
Spectrum Chemical Mfg Cor
P159150KGBL |
50 kg | Poly Pail |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
77-09-8 |
Specifications
77-09-8 | |
0.01 | |
Reagent | |
White | |
Phenolphthalein |
1 | |
98 to 101% | |
Poly Pail | |
125 g |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.