missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phenolphthalein, White, Powder, 98.0-101.0%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P159112KG
Specifications
Phenolphthalein | |
1 | |
98 to 101% | |
Poly Pail | |
12 kg |
77-09-8 | |
0.01 | |
Reagent | |
White |
Chemical Identifiers
77-09-8 |
CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.