missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 70%, ACS, 69.0-72.0%, Spectrum™ Chemical
$219.17 - $29109.32
Chemical Identifiers
CAS | 7601-90-3 |
---|---|
Molecular Formula | ClHO4 |
Molecular Weight (g/mol) | 100.45 |
MDL Number | MFCD00011325 |
InChI Key | VLTRZXGMWDSKGL-UHFFFAOYSA-N |
IUPAC Name | perchloric acid |
SMILES | O[Cl](=O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18-612-185
|
Spectrum Chemical Mfg Cor
P102525ML |
25 mL | Amber Glass Bottle |
Each for $219.17
|
|
||||
18-612-190
|
Spectrum Chemical Mfg Cor
P1025100ML |
100 mL | Amber Glass Bottle |
Each for $298.47
|
|
||||
18-612-186
|
Spectrum Chemical Mfg Cor
P1025500MLGL |
500 mL | Amber Glass Bottle |
Each for $858.21
|
|
||||
18-612-182
|
Spectrum Chemical Mfg Cor
P102525LTGL |
2.5 L | Glass Bottle |
Each for $2,092.18
|
|
||||
18-612-183
|
Spectrum Chemical Mfg Cor
P102525LTPV |
2.5 L | Poly-Coated Glass Bottle |
Each for $2,070.41
|
|
||||
18-612-184
|
Spectrum Chemical Mfg Cor
P1025200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
7601-90-3 | |
100.45 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
O[Cl](=O)(=O)=O |
ClHO4 | |
MFCD00011325 | |
perchloric acid |
Specifications
7601-90-3 | |
1 ppm | |
3e-05 | |
ClHO4 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
perchloric acid | |
69 to 72% |
1 | |
Amber Glass Bottle | |
25 mL | |
MFCD00011325 | |
O[Cl](=O)(=O)=O | |
100.45 | |
ACS |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.