missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perchloric Acid, 70%, ACS, 69.0-72.0%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor P102525LTPV
Specifications
7601-90-3 | |
1 ppm | |
3e-05 | |
ClHO4 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
perchloric acid | |
69 to 72% |
1 | |
Poly-Coated Glass Bottle | |
2.5 L | |
MFCD00011325 | |
O[Cl](=O)(=O)=O | |
100.45 | |
ACS |
Chemical Identifiers
7601-90-3 | |
100.45 | |
VLTRZXGMWDSKGL-UHFFFAOYSA-N | |
O[Cl](=O)(=O)=O |
ClHO4 | |
MFCD00011325 | |
perchloric acid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.