missing translation for 'onlineSavingsMsg'
Learn More

Oxalyl chloride, 2M soln. in dichloromethane

CAS: 79-37-8 | C2Cl2O2 | 126.92 g/mol

$115.27 - $247.14

Identifiants chimiques

CAS 79-37-8
Molecular Formula C2Cl2O2
Molecular Weight (g/mol) 126.92
MDL Number MFCD00000704
InChI Key CTSLXHKWHWQRSH-UHFFFAOYSA-N
Synonym oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
PubChem CID 65578
IUPAC Name oxalyl dichloride
SMILES C(=O)(C(=O)Cl)Cl
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAH31617AE
Afficher les documents
Thermo Scientific Chemicals
H31617AE
100 mL
chaque for $115.27
Il en reste null
 
AAH31617AP
Afficher les documents
Thermo Scientific Chemicals
H31617AP
500 mL
chaque for $247.14
Il en reste null
 
Description

Description

Reagent used in organic synthesis. Unncatalyzed reaction of silyl ketene acetals with oxalyl chloride yeilds symmetrical pulvinic acids. Catalytic syntheses of N-heterocyclic ynones and ynediones by in situ activation of carboxylic acids with oxalyl chloride. Oxalyl chloride was reportedly used in the first synthesis of dioxane tetraketone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reagent used in organic synthesis. Unncatalyzed reaction of silyl ketene acetals with oxalyl chloride yeilds symmetrical pulvinic acids. Catalytic syntheses of N-heterocyclic ynones and ynediones by in situ activation of carboxylic acids with oxalyl chloride. Oxalyl chloride was reportedly used in the first synthesis of dioxane tetraketone.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, light, bases, alkali metals, alcohols, dimethyl formamide.
Spécifications

Identifiants chimiques

79-37-8
126.92
CTSLXHKWHWQRSH-UHFFFAOYSA-N
65578
C(=O)(C(=O)Cl)Cl
C2Cl2O2
MFCD00000704
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
oxalyl dichloride

Spécifications

79-37-8
C2Cl2O2
100 mL
Moisture sensitive
Reacts with water.
C(=O)(C(=O)Cl)Cl
126.92
126.93
Oxalyl chloride
1.335
MFCD00000704
UN2922
oxalyl chloride, ethanedioyl dichloride, oxalic dichloride, oxaloyl chloride, oxalic acid dichloride, oxalic acid chloride, ethanedioyl chloride, oxalylchloride, unii-r4y96317dw, cocl 2
CTSLXHKWHWQRSH-UHFFFAOYSA-N
oxalyl dichloride
65578
2M soln. in dichloromethane
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H331-H314-H318-H351
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing cancer.

P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H332-H314-H350

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

missing translation for 'einecsNumber' : 201-200-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only