missing translation for 'onlineSavingsMsg'
Learn More

Watson Gloves Overtime Performance Gloves

Overtime Performance Gloves have a D3O™ knuckle bar that provides shock absorption and impact protection. Heavy duty microfibre palm with foam padding and silicone print provides a sure-grip.

Supplier:  Watson Gloves 025M

 View more versions of this product

Catalog No. 17-099-915


Only null left
Explore available promotions

Description

Description

  • D3O™ knuckle bar provides shock absorption and impact protection.
  • Spandex back made with WasteNot™ polyester yarn which is comprised of 96% recycled PET bottles
  • Heavy duty microfibre palm with foam padding
  • Silicone print provides a sure-grip
  • Air flow mesh on back of hand
  • Heavy duty rubber on fingers and thumb
  • Microfibre thumb saddle
  • Snug-fitting neoprene wrist
  • Secure Velcro™ closure
Specifications

Specifications

Spandex
EN388
Uncoated
Reusable
Ambidextrous
Spandex, Microfiber
Medium
Gunn
No
High Preformance, Mechanic, General Labor
Black
1 Pair of Gloves
No
Impact
Straight
23 cm
ANSI Impact 2
Industrial
Black
Partially Textured
Yes
Work Gloves
Non-sterile
EN388:3121XP
Full
Men's
Velcro™
No
Unlined
Spandex, Microfibre
9.5 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions